Learn More
MSC Vermont Gage Plug Gage, Tool Steel, 0.3335 in. Diameter, 2.0000 in. Length, 1 ea.
Catalog No. 5000511263
Encompass
This Class ZZ Plus plug gage is a high-precision tool steel instrument designed for Go inspection and for setting micrometers, verniers, and other measuring devices. It provides reliable and accurate measurements with a durable finish.
  • Diameter of 0.3335 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
  • Constructed from tool steel with a hardness of 60 to 65 Rc
  • Finish of 10 microinch or better
  • Round within 0.0001 in.
  • Includes certificate of accuracy with NIST traceability
  • Marked with size and direction of tolerance
Catalog No. 50-005-11263 Supplier MSC Supplier No. 88933353
This Class ZZ Plus plug gage is a high-precision tool steel instrument designed for Go inspection and for setting micrometers, verniers, and other measuring devices. It provides reliable and accurate measurements with a durable finish.
  • Diameter of 0.3335 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
  • Constructed from tool steel with a hardness of 60 to 65 Rc
  • Finish of 10 microinch or better
  • Round within 0.0001 in.
  • Includes certificate of accuracy with NIST traceability
  • Marked with size and direction of tolerance