Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.5800 in., Tool Steel, 1 ea.
Catalog No. 5000511191
Encompass
This Class ZZ Plus plug gage is designed for high-precision go inspection of holes and for setting measuring devices such as micrometers and verniers. Constructed from durable tool steel with a hardness of 60/65 Rc, it provides reliable performance in demanding measurement applications.
  • Diameter of 0.5800 in. with a tolerance of 0.000200 in.
  • Overall length of 2.0000 in.
  • Hardened tool steel construction for durability
  • Surface finish of 10 microinch or better
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in accordance with ASME B89.1 standards
Catalog No. 50-005-11191 Supplier MSC Supplier No. 88958004
This Class ZZ Plus plug gage is designed for high-precision go inspection of holes and for setting measuring devices such as micrometers and verniers. Constructed from durable tool steel with a hardness of 60/65 Rc, it provides reliable performance in demanding measurement applications.
  • Diameter of 0.5800 in. with a tolerance of 0.000200 in.
  • Overall length of 2.0000 in.
  • Hardened tool steel construction for durability
  • Surface finish of 10 microinch or better
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in accordance with ASME B89.1 standards