Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.8340 in., Tool Steel, 1 ea.
Catalog No. 5000511192
Encompass
This Class ZZ plug gage is designed for precision Go inspection, as well as for setting micrometers, verniers, and other measuring equipment. Constructed from durable tool steel, it offers high reliability for metrology applications.
  • Manufactured from tool steel with 60 to 65 Rc hardness
  • Meets ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Features 10 microinch surface finish
  • Round within 0.0001 in.
  • Tolerance of 0.0002 in.
  • Diameter is 0.8340 in. and overall length is 2.0000 in.
Catalog No. 50-005-11192 Supplier MSC Supplier No. 88983408
This Class ZZ plug gage is designed for precision Go inspection, as well as for setting micrometers, verniers, and other measuring equipment. Constructed from durable tool steel, it offers high reliability for metrology applications.
  • Manufactured from tool steel with 60 to 65 Rc hardness
  • Meets ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Features 10 microinch surface finish
  • Round within 0.0001 in.
  • Tolerance of 0.0002 in.
  • Diameter is 0.8340 in. and overall length is 2.0000 in.