Learn More
MSC Vermont Gage Class ZZ Plug Gage, Tool Steel, 0.8470 in. Diameter, 2 in. Length, 1 ea
Catalog No. 5000511193
Encompass
This Class ZZ plug gage is designed for precision Go inspection, setting micrometers, and calibrating various measuring devices. Constructed from high-quality tool steel, this gage provides durable performance and reliable accuracy for industrial inspection tasks.
  • Tool steel construction with 60/65 Rc hardness for long-lasting durability
  • Features a 0.8470 in. diameter and 2 in. overall length
  • Tolerance of +.0002 / -.0000 in.
  • Finished to 10 microinch or better for smooth surface contact
  • Roundness tolerance within .0001 in.
  • Includes a certificate of accuracy with NIST traceability
  • Marked with size and direction of tolerance for easy identification
Catalog No. 50-005-11193 Supplier MSC Supplier No. 88984703
This Class ZZ plug gage is designed for precision Go inspection, setting micrometers, and calibrating various measuring devices. Constructed from high-quality tool steel, this gage provides durable performance and reliable accuracy for industrial inspection tasks.
  • Tool steel construction with 60/65 Rc hardness for long-lasting durability
  • Features a 0.8470 in. diameter and 2 in. overall length
  • Tolerance of +.0002 / -.0000 in.
  • Finished to 10 microinch or better for smooth surface contact
  • Roundness tolerance within .0001 in.
  • Includes a certificate of accuracy with NIST traceability
  • Marked with size and direction of tolerance for easy identification