Learn More
MSC Vermont Gage Class ZZ Plus Pin Gage, Tool Steel, 0.1870 in. Diameter, 2.0000 in. Length, 1 ea.
Catalog No. 5000510463
Encompass
This Class ZZ Plus pin gage is designed for precision inspection tasks and serves as a reliable standard for checking measuring instruments. Featuring a 0.1870 in. diameter and a 2.0000 in. overall length, this gage is constructed from durable, heat-treated tool steel. It provides accurate, repeatable results for go inspection and the setting of micrometers or verniers.
  • Manufactured from 52100 tool steel with a hardness of 60/65 Rc
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microinch surface finish or better
  • Tolerance of 0.0000 in. to +0.0002 in.
  • Constructed to ASME B89.1 standards
  • Marked with size and direction of tolerance for identification
Catalog No. 50-005-10463 Supplier MSC Supplier No. 88918701
This Class ZZ Plus pin gage is designed for precision inspection tasks and serves as a reliable standard for checking measuring instruments. Featuring a 0.1870 in. diameter and a 2.0000 in. overall length, this gage is constructed from durable, heat-treated tool steel. It provides accurate, repeatable results for go inspection and the setting of micrometers or verniers.
  • Manufactured from 52100 tool steel with a hardness of 60/65 Rc
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microinch surface finish or better
  • Tolerance of 0.0000 in. to +0.0002 in.
  • Constructed to ASME B89.1 standards
  • Marked with size and direction of tolerance for identification