Learn More
MSC Vermont Gage Class ZZ Plus Pin Gage, 0.2530 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000510432
Encompass
This Class ZZ plug gage is designed for precise measurement and inspection tasks, featuring a 0.2530 in. diameter and a 2.0000 in. overall length. It is constructed from durable tool steel and engineered for applications including go inspection and the setting of micrometers, verniers, and other measuring devices.
  • Constructed from tool steel for enhanced durability
  • Finished to 10 micro or better for consistent accuracy
  • Includes a certificate of accuracy with NIST traceability
  • Built to meet ASME B89.1 standards
  • Reversible style for extended service life
  • Tolerance of +0.0002 / -0.0000 in.
Catalog No. 50-005-10432 Supplier MSC Supplier No. 88925300
This Class ZZ plug gage is designed for precise measurement and inspection tasks, featuring a 0.2530 in. diameter and a 2.0000 in. overall length. It is constructed from durable tool steel and engineered for applications including go inspection and the setting of micrometers, verniers, and other measuring devices.
  • Constructed from tool steel for enhanced durability
  • Finished to 10 micro or better for consistent accuracy
  • Includes a certificate of accuracy with NIST traceability
  • Built to meet ASME B89.1 standards
  • Reversible style for extended service life
  • Tolerance of +0.0002 / -0.0000 in.