Learn More
MSC Vermont Gage Class ZZ Pin Gage, Tool Steel, 0.3225 in. Diameter, 2.0000 in. Length, 1 ea
Catalog No. 5000510439
Encompass
This Class ZZ plug gage is designed for precise measurement and inspection applications, including go inspection and the setting of micrometers, verniers, and other measuring devices. Constructed from 52100 tool steel, the gage is heat-treated to a hardness of 60 to 62 RC for durability and wear resistance.
  • Manufactured from high-quality 52100 tool steel
  • Heat-treated to 60 to 62 RC for extended life
  • Provides accurate measurement with a 0.0002 in. tolerance
  • Features a surface finish of 10 microinch or better
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-10439 Supplier MSC Supplier No. 88932256
This Class ZZ plug gage is designed for precise measurement and inspection applications, including go inspection and the setting of micrometers, verniers, and other measuring devices. Constructed from 52100 tool steel, the gage is heat-treated to a hardness of 60 to 62 RC for durability and wear resistance.
  • Manufactured from high-quality 52100 tool steel
  • Heat-treated to 60 to 62 RC for extended life
  • Provides accurate measurement with a 0.0002 in. tolerance
  • Features a surface finish of 10 microinch or better
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards