Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, 1.0000 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000510396
Encompass
This Class ZZ steel plug and pin gage is designed for precision inspection tasks, including NoGo measurements and the setting of micrometers, verniers, and other measuring devices. It features a 1.0000 in. diameter and an overall length of 2 in.
  • Tolerance of 0.00020 in.
  • Hardness rating of RC 60/65
  • Surface finish of 10 microinch or better
  • Cylindrical shape
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025:2005 standards by A2LA
Catalog No. 50-005-10396 Supplier MSC Supplier No. 89099998
This Class ZZ steel plug and pin gage is designed for precision inspection tasks, including NoGo measurements and the setting of micrometers, verniers, and other measuring devices. It features a 1.0000 in. diameter and an overall length of 2 in.
  • Tolerance of 0.00020 in.
  • Hardness rating of RC 60/65
  • Surface finish of 10 microinch or better
  • Cylindrical shape
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025:2005 standards by A2LA