Learn More
MSC Vermont Gage Class ZZ Gage Pin, Tool Steel, 0.4965 in. Diameter, 2 in. Length, 1 ea
Catalog No. 5000510362
Encompass
Class ZZ gage pins provide a reliable solution for precise dimensional measurements and quality control inspections. These pins are designed to facilitate accurate hole diameter assessment, slot width determination, and the setting of various measuring instruments such as micrometers and verniers. Constructed from durable tool steel, they support both Go and NoGo inspection applications.
  • Constructed from tool steel with 60/65 Rc hardness
  • Features a 10 microinch finish for smooth operation
  • Maintains roundness within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability
  • Diameter of 0.4965 in. with a tolerance of 0.0002 in.
  • Overall length of 2 in.
Catalog No. 50-005-10362 Supplier MSC Supplier No. 89049654
Class ZZ gage pins provide a reliable solution for precise dimensional measurements and quality control inspections. These pins are designed to facilitate accurate hole diameter assessment, slot width determination, and the setting of various measuring instruments such as micrometers and verniers. Constructed from durable tool steel, they support both Go and NoGo inspection applications.
  • Constructed from tool steel with 60/65 Rc hardness
  • Features a 10 microinch finish for smooth operation
  • Maintains roundness within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability
  • Diameter of 0.4965 in. with a tolerance of 0.0002 in.
  • Overall length of 2 in.