Learn More
MSC Vermont Gage Class ZZ Pin Gage, 0.6580 in. Diameter, Tool Steel, 2 in. Length, 1 ea.
Catalog No. 5000516433
Encompass
This Class ZZ pin gage is designed for high-precision measurement applications, including setting micrometers and conducting inspections with verniers and other measuring instruments. It is constructed from heat-treated tool steel for durability and is centerless lapped to ensure high accuracy.
  • Tolerance of +.0000/-.0002 in.
  • Overall length of 2 in.
  • Diameter of 0.6580 in.
  • Hardness of 60 to 65 Rc
  • Finish of 10 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Size and direction of tolerance marked on the gage
Catalog No. 50-005-16433 Supplier MSC Supplier No. 89065809
This Class ZZ pin gage is designed for high-precision measurement applications, including setting micrometers and conducting inspections with verniers and other measuring instruments. It is constructed from heat-treated tool steel for durability and is centerless lapped to ensure high accuracy.
  • Tolerance of +.0000/-.0002 in.
  • Overall length of 2 in.
  • Diameter of 0.6580 in.
  • Hardness of 60 to 65 Rc
  • Finish of 10 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Size and direction of tolerance marked on the gage