Learn More
MSC Vermont Gage Class ZZ Pin Gage, 0.1600 in. Diameter, Plus Tolerance, 2.0000 in. OAL, 1 ea.
Catalog No. 5000516442
Encompass
This Class ZZ plug gage is designed for precise measurement applications, including go inspection and the setting of micrometers, verniers, and other measuring devices. Constructed from high-quality tool steel, the gage offers durability and reliable performance in metrology environments.
  • Hardened to 60-62 RC for enhanced wear resistance
  • Manufactured with a 0.000200 in. tolerance
  • Features a 10 μin. finish for smooth contact
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Dimensions include a 0.1600 in. diameter and 2.0000 in. overall length
Catalog No. 50-005-16442 Supplier MSC Supplier No. 88916002
This Class ZZ plug gage is designed for precise measurement applications, including go inspection and the setting of micrometers, verniers, and other measuring devices. Constructed from high-quality tool steel, the gage offers durability and reliable performance in metrology environments.
  • Hardened to 60-62 RC for enhanced wear resistance
  • Manufactured with a 0.000200 in. tolerance
  • Features a 10 μin. finish for smooth contact
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Dimensions include a 0.1600 in. diameter and 2.0000 in. overall length