Learn More
MSC Vermont Gage Class ZZ Pin Gage, Minus Tolerance, 0.4145 in., 2 in. Length, 1 ea.
Catalog No. 5000516282
Encompass
This Class ZZ pin gage is a precision-engineered tool designed for accurate hole diameter measurement and inspection. Constructed from high-quality 52100 tool steel, this 0.4145 in. diameter plug gage offers high durability and wear resistance in demanding measurement applications. It is centerless lapped with a 10 micro-finish and heat-treated to a hardness of 60 to 65 HRC for long-term stability and corrosion resistance.
  • NIST traceable with included certificate of accuracy
  • Minus tolerance of -0.0002/+0.0000 in.
  • Overall length of 2 in.
  • Electro-etched size marking for easy identification
  • Suitable for checking hole locations, slot widths, and setting micrometers or calipers
Catalog No. 50-005-16282 Supplier MSC Supplier No. 89041453
This Class ZZ pin gage is a precision-engineered tool designed for accurate hole diameter measurement and inspection. Constructed from high-quality 52100 tool steel, this 0.4145 in. diameter plug gage offers high durability and wear resistance in demanding measurement applications. It is centerless lapped with a 10 micro-finish and heat-treated to a hardness of 60 to 65 HRC for long-term stability and corrosion resistance.
  • NIST traceable with included certificate of accuracy
  • Minus tolerance of -0.0002/+0.0000 in.
  • Overall length of 2 in.
  • Electro-etched size marking for easy identification
  • Suitable for checking hole locations, slot widths, and setting micrometers or calipers