Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 0.0430 in., Class ZZ, 1 ea.
Catalog No. 5000516240
Encompass
The Class ZZ pin gage is a high-precision tool designed for measuring hole sizes and calibrating instruments such as micrometers and verniers. Constructed from durable 52100 tool steel and heat-treated to 60/65 Rc, this gage offers long-lasting performance and wear resistance. It features a centerless lapped finish and ground, smooth edges to ensure measurement accuracy and ease of use in industrial inspection environments.
  • Diameter of 0.0430 in. with a tolerance of +0.0002/-.0000 in.
  • Overall length of 2 in.
  • Hardened tool steel construction with a 10 microinch finish for superior precision
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-16240 Supplier MSC Supplier No. 88904305
The Class ZZ pin gage is a high-precision tool designed for measuring hole sizes and calibrating instruments such as micrometers and verniers. Constructed from durable 52100 tool steel and heat-treated to 60/65 Rc, this gage offers long-lasting performance and wear resistance. It features a centerless lapped finish and ground, smooth edges to ensure measurement accuracy and ease of use in industrial inspection environments.
  • Diameter of 0.0430 in. with a tolerance of +0.0002/-.0000 in.
  • Overall length of 2 in.
  • Hardened tool steel construction with a 10 microinch finish for superior precision
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards