Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Tool Steel, 0.1395 in. Dia., Plus, 1 ea.
Catalog No. 5000516096
Encompass
This Class ZZ plus plug gage is a precision measurement tool designed for tasks such as hole size verification, distance calculation between holes, and checking slot widths. Engineered for reliability, this gage serves as an economical solution for high-accuracy Go gaging applications in industrial inspection and metrology.
  • Diameter of 0.1395 in.
  • Tolerance of +0.0002 in.
  • Overall length of 2.0000 in.
  • Constructed from durable 52100 tool steel, heat-treated to 60 to 62 RC for enhanced wear resistance.
  • Surface finished to 10 micro-finish or better for smooth operation.
  • Meets ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.
  • Reversible design for increased utility.
Catalog No. 50-005-16096 Supplier MSC Supplier No. 88913959
This Class ZZ plus plug gage is a precision measurement tool designed for tasks such as hole size verification, distance calculation between holes, and checking slot widths. Engineered for reliability, this gage serves as an economical solution for high-accuracy Go gaging applications in industrial inspection and metrology.
  • Diameter of 0.1395 in.
  • Tolerance of +0.0002 in.
  • Overall length of 2.0000 in.
  • Constructed from durable 52100 tool steel, heat-treated to 60 to 62 RC for enhanced wear resistance.
  • Surface finished to 10 micro-finish or better for smooth operation.
  • Meets ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.
  • Reversible design for increased utility.