Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, Steel, 0.4730 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000516009
Encompass
This Class ZZ minus plug and pin gage is a precision measurement tool with a diameter of 0.4730 in. and an overall length of 2 in. Designed for durability and accuracy, it is suitable for setting micrometers, verniers, and other measuring devices, as well as for conducting NoGo inspections.
  • Constructed from steel with a 10 microinch finish
  • Features a minus tolerance direction with a tolerance of 0.000200 in.
  • Includes a certificate of accuracy with NIST traceability
  • Reversible style design
  • Size and direction of tolerance marked on the gauge
Catalog No. 50-005-16009 Supplier MSC Supplier No. 89047302
This Class ZZ minus plug and pin gage is a precision measurement tool with a diameter of 0.4730 in. and an overall length of 2 in. Designed for durability and accuracy, it is suitable for setting micrometers, verniers, and other measuring devices, as well as for conducting NoGo inspections.
  • Constructed from steel with a 10 microinch finish
  • Features a minus tolerance direction with a tolerance of 0.000200 in.
  • Includes a certificate of accuracy with NIST traceability
  • Reversible style design
  • Size and direction of tolerance marked on the gauge