Learn More
MSC Vermont Gage Plug Gage, Class ZZ, Tool Steel, 0.2605 in. Diameter, 1 ea.
Catalog No. 5000515966
Encompass
This Class ZZ plug gage is designed for precision inspection tasks, including go inspection and the setting of micrometers, verniers, and other measuring devices. Constructed from high-quality tool steel, it offers durability and consistent measurement accuracy in demanding workshop environments.
  • Manufactured from tool steel with a hardness of 60/62 Rc
  • Provides a tolerance of 0.0002 in.
  • Features a finish of 10 microinch or better
  • Meets ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Measures 2 in. in overall length
  • Round within 0.0001 in.
Catalog No. 50-005-15966 Supplier MSC Supplier No. 88926050
This Class ZZ plug gage is designed for precision inspection tasks, including go inspection and the setting of micrometers, verniers, and other measuring devices. Constructed from high-quality tool steel, it offers durability and consistent measurement accuracy in demanding workshop environments.
  • Manufactured from tool steel with a hardness of 60/62 Rc
  • Provides a tolerance of 0.0002 in.
  • Features a finish of 10 microinch or better
  • Meets ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Measures 2 in. in overall length
  • Round within 0.0001 in.