Learn More
MSC Vermont Gage Class ZZ Plus Pin Gage, Tool Steel, 0.3935 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000515808
Encompass
This Class ZZ plug gage is designed for precise measurement applications, including go inspections, setting micrometers, and calibrating verniers or other measuring devices. Constructed from high-quality 52100 tool steel, the gage offers durability and consistency for industrial inspection tasks.
  • Features a diameter of 0.3935 in. and an overall length of 2.0000 in.
  • Tolerance of 0.0000/+0.0002 in. ensures high precision.
  • Heat-treated to a hardness of 60 to 62 RC for enhanced wear resistance.
  • Includes a certificate of accuracy with NIST traceability to verify compliance with ASME B89.1 standards.
  • Surface finish of 10 micro-inches for smooth operation.
Catalog No. 50-005-15808 Supplier MSC Supplier No. 88939350
This Class ZZ plug gage is designed for precise measurement applications, including go inspections, setting micrometers, and calibrating verniers or other measuring devices. Constructed from high-quality 52100 tool steel, the gage offers durability and consistency for industrial inspection tasks.
  • Features a diameter of 0.3935 in. and an overall length of 2.0000 in.
  • Tolerance of 0.0000/+0.0002 in. ensures high precision.
  • Heat-treated to a hardness of 60 to 62 RC for enhanced wear resistance.
  • Includes a certificate of accuracy with NIST traceability to verify compliance with ASME B89.1 standards.
  • Surface finish of 10 micro-inches for smooth operation.