Learn More
MSC Vermont Gage Individual Pin Gages, Tool Steel, Class ZZ, 2 in., 1 ea.
Catalog No. 5000515789
Encompass
These Class ZZ gage pins are designed for precision measurements, including hole diameters, slot widths, and calculating distances between holes. They serve as reliable tools for Go/NoGo inspections and for verifying the accuracy of micrometers and verniers. These pins are ideal for maintaining calibration compliance in various metrology applications.
  • Constructed from durable tool steel
  • Tolerance of 0.0002 in. (0.005 mm)
  • Hardness rating of 60/65 Rc
  • Surface finish of 10 microinch or better
  • Roundness within 0.0001 in. (0.0025 mm)
  • Available in Go (plus) or NoGo (minus) tolerance
  • NIST traceable with included certificate of accuracy
Catalog No. 50-005-15789 Supplier MSC Supplier No. 88988803
These Class ZZ gage pins are designed for precision measurements, including hole diameters, slot widths, and calculating distances between holes. They serve as reliable tools for Go/NoGo inspections and for verifying the accuracy of micrometers and verniers. These pins are ideal for maintaining calibration compliance in various metrology applications.
  • Constructed from durable tool steel
  • Tolerance of 0.0002 in. (0.005 mm)
  • Hardness rating of 60/65 Rc
  • Surface finish of 10 microinch or better
  • Roundness within 0.0001 in. (0.0025 mm)
  • Available in Go (plus) or NoGo (minus) tolerance
  • NIST traceable with included certificate of accuracy