Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.4890 in., 2 in. Length, 1 ea.
Catalog No. 5000515756
Encompass
These Class ZZ Plus plug gages are designed for general-purpose measurement tasks, including hole size verification, hole distance calculation, and slot width gaging. Constructed from tool steel with a 60/65 Rc hardness, these gages provide durable wear resistance for go inspection and setting of micrometers and verniers.
  • Diameter of 0.4890 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in. (0.00508 mm)
  • Finished to 10 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Constructed to ASME B89.1 standards
Catalog No. 50-005-15756 Supplier MSC Supplier No. 88948906
These Class ZZ Plus plug gages are designed for general-purpose measurement tasks, including hole size verification, hole distance calculation, and slot width gaging. Constructed from tool steel with a 60/65 Rc hardness, these gages provide durable wear resistance for go inspection and setting of micrometers and verniers.
  • Diameter of 0.4890 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in. (0.00508 mm)
  • Finished to 10 microinch or better
  • Includes certificate of accuracy with NIST traceability
  • Constructed to ASME B89.1 standards