Learn More
MSC Vermont Gage Plug Gage, Class ZZ Minus, Steel, 0.2965 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000515726
Encompass
The class ZZ minus plug and pin gage is a precision measuring instrument designed for checking hole sizes, setting micrometers, and calibrating measuring devices. Constructed from 52100 tool steel, this gage provides durability and accuracy for industrial inspection applications.
  • Diameter of 0.2965 in. with 2 in. overall length
  • Heat-treated to 60 to 62 RC for enhanced wear resistance
  • Features a 10 micro finish
  • Centerless lapped with well-ground ends and smooth edges
  • Minus tolerance of -0.0002 in. to +0.0000 in.
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-15726 Supplier MSC Supplier No. 89029656
The class ZZ minus plug and pin gage is a precision measuring instrument designed for checking hole sizes, setting micrometers, and calibrating measuring devices. Constructed from 52100 tool steel, this gage provides durability and accuracy for industrial inspection applications.
  • Diameter of 0.2965 in. with 2 in. overall length
  • Heat-treated to 60 to 62 RC for enhanced wear resistance
  • Features a 10 micro finish
  • Centerless lapped with well-ground ends and smooth edges
  • Minus tolerance of -0.0002 in. to +0.0000 in.
  • Includes certificate of accuracy with NIST traceability