Learn More
MSC Vermont Gage Pin Gage, Class ZZ Minus, 0.3685 in. Diameter, Steel, 1 ea.
Catalog No. 5000515728
Encompass
This Class ZZ minus plug and pin gage is a precision measurement tool designed for inspecting components and calibrating measuring devices. With a diameter of 0.3685 in., it provides high-accuracy dimensional verification in various industrial applications.
  • Diameter of 0.3685 in.
  • Constructed from steel for durability
  • Features a 10 microfinish coating
  • Designed for use as a NoGo gauge, setting micrometers, or calibrating verniers
  • Includes a certificate of accuracy with NIST traceability
  • Overall length of 2 in.
  • Tolerance of 0.000200 in.
  • Reversible style
Catalog No. 50-005-15728 Supplier MSC Supplier No. 89036859
This Class ZZ minus plug and pin gage is a precision measurement tool designed for inspecting components and calibrating measuring devices. With a diameter of 0.3685 in., it provides high-accuracy dimensional verification in various industrial applications.
  • Diameter of 0.3685 in.
  • Constructed from steel for durability
  • Features a 10 microfinish coating
  • Designed for use as a NoGo gauge, setting micrometers, or calibrating verniers
  • Includes a certificate of accuracy with NIST traceability
  • Overall length of 2 in.
  • Tolerance of 0.000200 in.
  • Reversible style