Learn More
MSC Vermont Gage Class ZZ Pin Gage, 0.9440 in. Diameter, Tool Steel, 2 in. Length, 1 ea.
Catalog No. 5000515712
Encompass
This Class ZZ plug gage is designed for precise measurement of hole sizes and is suitable for use in setting micrometers, verniers, and other measuring instruments. Constructed from 52100 tool steel and heat-treated to 60 to 62 RC, this gage offers durability and high-precision inspection capabilities.
  • Manufactured from 52100 tool steel
  • Tolerance of +0.0002 in./-0.0000 in.
  • Overall length of 2.0000 in.
  • Diameter of 0.9440 in.
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Finish of 10 micro or better
  • Reversible style
Catalog No. 50-005-15712 Supplier MSC Supplier No. 88994405
This Class ZZ plug gage is designed for precise measurement of hole sizes and is suitable for use in setting micrometers, verniers, and other measuring instruments. Constructed from 52100 tool steel and heat-treated to 60 to 62 RC, this gage offers durability and high-precision inspection capabilities.
  • Manufactured from 52100 tool steel
  • Tolerance of +0.0002 in./-0.0000 in.
  • Overall length of 2.0000 in.
  • Diameter of 0.9440 in.
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Finish of 10 micro or better
  • Reversible style