Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Tool Steel, 0.4815 in. Diameter, 2.0000 in. Length, 1 ea
Catalog No. 5000515647
Encompass
This Class ZZ plug gage is designed for precision Go inspection, setting micrometers, and calibrating various measuring devices. Crafted from durable tool steel and heat-treated for longevity, this gage provides accurate measurement control in demanding industrial environments.
  • Hardness of 60 to 65 Rc
  • Finish of 10 microinch
  • Round within .0001 in.
  • Tolerance of .0002 in.
  • Overall length of 2.0000 in.
  • Diameter of 0.4815 in.
  • Includes certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Marked with diameter and direction of tolerance
Catalog No. 50-005-15647 Supplier MSC Supplier No. 88948153
This Class ZZ plug gage is designed for precision Go inspection, setting micrometers, and calibrating various measuring devices. Crafted from durable tool steel and heat-treated for longevity, this gage provides accurate measurement control in demanding industrial environments.
  • Hardness of 60 to 65 Rc
  • Finish of 10 microinch
  • Round within .0001 in.
  • Tolerance of .0002 in.
  • Overall length of 2.0000 in.
  • Diameter of 0.4815 in.
  • Includes certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Marked with diameter and direction of tolerance