Learn More
MSC Vermont Gage Class ZZ Pin Gage, Tool Steel, 0.3185 in. Diameter, 2.0000 in. Length, 1 ea.
Catalog No. 5000515643
Encompass
This Class ZZ plus plug gage is a high-precision measurement tool designed for Go inspection and for setting micrometers, verniers, and other measuring devices. Engineered from 52100 tool steel and heat-treated to 60 to 62 RC, this gage provides durability and dimensional stability for critical metrology applications.
  • Diameter of 0.3185 in. with a plus tolerance of 0.0002 in.
  • Overall length of 2.0000 in.
  • Centerless lapped for a 10 microfinish or better
  • Round within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in the United States of America
  • Compliant with ASME B89.1 standards
Catalog No. 50-005-15643 Supplier MSC Supplier No. 88931852
This Class ZZ plus plug gage is a high-precision measurement tool designed for Go inspection and for setting micrometers, verniers, and other measuring devices. Engineered from 52100 tool steel and heat-treated to 60 to 62 RC, this gage provides durability and dimensional stability for critical metrology applications.
  • Diameter of 0.3185 in. with a plus tolerance of 0.0002 in.
  • Overall length of 2.0000 in.
  • Centerless lapped for a 10 microfinish or better
  • Round within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in the United States of America
  • Compliant with ASME B89.1 standards