Learn More
MSC Vermont Gage Class ZZ Pin Gage, 0.7510 in. Diameter, Steel, 1 ea.
Catalog No. 5000515560
Encompass
This Class ZZ steel pin gage is designed for high-precision measurement and inspection applications, particularly as a NoGo gage. Engineered for durability and reliability, it features a hardened construction to ensure consistent performance in demanding workshop environments.
  • Diameter of 0.7510 in.
  • Overall length of 2 in.
  • Tolerance of 0.000200 in.
  • Minus tolerance direction for NoGo inspection
  • Surface finish of 10 microinch or better
  • Heat-treated to 60 to 62 Rc for wear resistance
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-15560 Supplier MSC Supplier No. 89075105
This Class ZZ steel pin gage is designed for high-precision measurement and inspection applications, particularly as a NoGo gage. Engineered for durability and reliability, it features a hardened construction to ensure consistent performance in demanding workshop environments.
  • Diameter of 0.7510 in.
  • Overall length of 2 in.
  • Tolerance of 0.000200 in.
  • Minus tolerance direction for NoGo inspection
  • Surface finish of 10 microinch or better
  • Heat-treated to 60 to 62 Rc for wear resistance
  • Includes certificate of accuracy with NIST traceability