Learn More
MSC Vermont Gage Class ZZ Pin Gage, 0.2985 in. Diameter, 2 in. Length, Tool Steel, 1 ea.
Catalog No. 5000515529
Encompass
This Class ZZ minus plug and pin gage is designed for precise measurement applications, including inspecting hole diameters, checking slot widths, and setting micrometers or vernier calipers. Manufactured from 52100 tool steel, this gage provides high durability and accurate performance for Go/NoGo inspection tasks.
  • Diameter of 0.2985 in. with a minus tolerance of 0.0002 in.
  • Overall length of 2 in.
  • Hardness rated at 60 to 62 Rc.
  • Finish of 10 microinch for smooth surface quality.
  • Roundness within 0.0001 in.
  • Includes certificate of accuracy with NIST traceability.
  • Marked with diameter and direction of tolerance.
Catalog No. 50-005-15529 Supplier MSC Supplier No. 89029854
This Class ZZ minus plug and pin gage is designed for precise measurement applications, including inspecting hole diameters, checking slot widths, and setting micrometers or vernier calipers. Manufactured from 52100 tool steel, this gage provides high durability and accurate performance for Go/NoGo inspection tasks.
  • Diameter of 0.2985 in. with a minus tolerance of 0.0002 in.
  • Overall length of 2 in.
  • Hardness rated at 60 to 62 Rc.
  • Finish of 10 microinch for smooth surface quality.
  • Roundness within 0.0001 in.
  • Includes certificate of accuracy with NIST traceability.
  • Marked with diameter and direction of tolerance.