Learn More
MSC Vermont Gage Pin Gage, Class ZZ, 0.6840 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000515558
Encompass
This Class ZZ minus plug gage is a precision-engineered tool designed for high-accuracy inspection and calibration tasks. Measuring 0.6840 in. in diameter with a tolerance of 0.0002 in., it is ideal for verifying hole sizes, checking go/no-go dimensions, and calibrating measuring devices like micrometers and verniers. Each gage is constructed from durable tool steel for long-lasting performance in industrial environments.
  • Diameter of 0.6840 in. with a class ZZ tolerance of 0.0002 in.
  • Overall length of 2 in.
  • Hardness rating of 60/65 Rc for wear resistance.
  • Surface finish of 10 microinch for smooth measurement insertion.
  • Includes a certificate of accuracy with NIST traceability.
  • Marked with diameter and direction of tolerance for easy identification.
  • Round within 0.0001 in.
Catalog No. 50-005-15558 Supplier MSC Supplier No. 89068407
This Class ZZ minus plug gage is a precision-engineered tool designed for high-accuracy inspection and calibration tasks. Measuring 0.6840 in. in diameter with a tolerance of 0.0002 in., it is ideal for verifying hole sizes, checking go/no-go dimensions, and calibrating measuring devices like micrometers and verniers. Each gage is constructed from durable tool steel for long-lasting performance in industrial environments.
  • Diameter of 0.6840 in. with a class ZZ tolerance of 0.0002 in.
  • Overall length of 2 in.
  • Hardness rating of 60/65 Rc for wear resistance.
  • Surface finish of 10 microinch for smooth measurement insertion.
  • Includes a certificate of accuracy with NIST traceability.
  • Marked with diameter and direction of tolerance for easy identification.
  • Round within 0.0001 in.