Learn More
MSC Vermont Gage Class ZZ Pin Gage, 0.4975 in. Diameter, 52100 Tool Steel, 2 in. Length, 1 ea.
Catalog No. 5000515426
Encompass
This Class ZZ plug gage is designed for precision inspection applications, including checking micrometers, verniers, and other measuring devices. Constructed from 52100 tool steel and heat-treated to 60 to 62 RC, this gage offers durability and high-precision performance.
  • Diameter of 0.4975 in.
  • Overall length of 2.0000 in.
  • Tolerance of +0.0002 in. / -0.0000 in.
  • Class ZZ tolerance rating per ASME B89.1
  • High-quality finish of 10 micro or better
  • Includes certificate of accuracy with NIST traceability
  • Constructed from durable 52100 tool steel
Catalog No. 50-005-15426 Supplier MSC Supplier No. 88949755
This Class ZZ plug gage is designed for precision inspection applications, including checking micrometers, verniers, and other measuring devices. Constructed from 52100 tool steel and heat-treated to 60 to 62 RC, this gage offers durability and high-precision performance.
  • Diameter of 0.4975 in.
  • Overall length of 2.0000 in.
  • Tolerance of +0.0002 in. / -0.0000 in.
  • Class ZZ tolerance rating per ASME B89.1
  • High-quality finish of 10 micro or better
  • Includes certificate of accuracy with NIST traceability
  • Constructed from durable 52100 tool steel