Learn More
MSC Vermont Gage Class ZZ Plus Pin Gage, Tool Steel, 0.4870 in. Diameter, 2.0000 in. Length, 1 ea.
Catalog No. 5000515149
Encompass
This Class ZZ plug gage is designed for precision inspection and measurement applications. It is used for go inspection, setting micrometers, calibrating verniers, and verifying various other measuring instruments. Constructed from 52100 tool steel and heat-treated for durability, this gage provides reliable performance in industrial environments.
  • Diameter of 0.4870 in. with a tolerance of +0.0002 in./-0.0000 in.
  • Overall length of 2.0000 in.
  • Manufactured from 52100 tool steel, heat-treated to 60 to 65 Rc hardness.
  • Surface finish of 10 micro-inches or better.
  • Roundness within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-15149 Supplier MSC Supplier No. 88948708
This Class ZZ plug gage is designed for precision inspection and measurement applications. It is used for go inspection, setting micrometers, calibrating verniers, and verifying various other measuring instruments. Constructed from 52100 tool steel and heat-treated for durability, this gage provides reliable performance in industrial environments.
  • Diameter of 0.4870 in. with a tolerance of +0.0002 in./-0.0000 in.
  • Overall length of 2.0000 in.
  • Manufactured from 52100 tool steel, heat-treated to 60 to 65 Rc hardness.
  • Surface finish of 10 micro-inches or better.
  • Roundness within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.