Learn More
MSC Vermont Gage Class ZZ Pin Gage, Tool Steel, 0.4090 in. Diameter, 2.0000 in. OAL, 1 ea
Catalog No. 5000515071
Encompass
This Class ZZ plug gage is designed for precision Go inspection, setting micrometers, and calibrating various measuring devices. Constructed from high-quality tool steel, it offers durability and consistent measurement accuracy for demanding shop floor or laboratory environments.
  • Diameter of 0.4090 in. with a tolerance of 0.000200 in.
  • Overall length of 2.0000 in.
  • Heat-treated to 60 to 62 RC for enhanced hardness and longevity.
  • Surface finish of 10 micro or better for smooth operation.
  • Meets ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-15071 Supplier MSC Supplier No. 88940903
This Class ZZ plug gage is designed for precision Go inspection, setting micrometers, and calibrating various measuring devices. Constructed from high-quality tool steel, it offers durability and consistent measurement accuracy for demanding shop floor or laboratory environments.
  • Diameter of 0.4090 in. with a tolerance of 0.000200 in.
  • Overall length of 2.0000 in.
  • Heat-treated to 60 to 62 RC for enhanced hardness and longevity.
  • Surface finish of 10 micro or better for smooth operation.
  • Meets ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.