Learn More
MSC Vermont Gage Class ZZ Plug Gage, 0.4695 in. Diameter, Steel, 2.0000 in. Length, 1 ea.
Catalog No. 5000515072
Encompass
This Class ZZ plug gage is designed for precise measurement applications, including go inspection and the calibration of micrometers, verniers, and other measuring instruments. Constructed from 52100 tool steel, this reversible gage is heat-treated to 60 to 62 RC for enhanced durability and performance.
  • Diameter of 0.4695 in. with a tolerance of 0.0000 to +0.0002 in.
  • Overall length of 2.0000 in.
  • Finish of 10 micro to ensure smooth operation.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured in the United States according to ASME B89.1 standards.
Catalog No. 50-005-15072 Supplier MSC Supplier No. 88946959
This Class ZZ plug gage is designed for precise measurement applications, including go inspection and the calibration of micrometers, verniers, and other measuring instruments. Constructed from 52100 tool steel, this reversible gage is heat-treated to 60 to 62 RC for enhanced durability and performance.
  • Diameter of 0.4695 in. with a tolerance of 0.0000 to +0.0002 in.
  • Overall length of 2.0000 in.
  • Finish of 10 micro to ensure smooth operation.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured in the United States according to ASME B89.1 standards.