Learn More
MSC Vermont Gage Pin Gage, Class ZZ, 0.0225 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000513985
Encompass
This Class ZZ plug and pin gage is designed for precision inspection and calibration tasks. Manufactured from tool steel with a hardness of Rc 60/65, this gage provides durable performance for setting micrometers, checking verniers, and other measurement device applications.
  • Diameter of 0.0225 in.
  • Overall length of 2 in.
  • Constructed from tool steel (Rc 60/65)
  • Finish of 10 micro or better
  • Minus tolerance direction
  • Reversible style
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-13985 Supplier MSC Supplier No. 89002257
This Class ZZ plug and pin gage is designed for precision inspection and calibration tasks. Manufactured from tool steel with a hardness of Rc 60/65, this gage provides durable performance for setting micrometers, checking verniers, and other measurement device applications.
  • Diameter of 0.0225 in.
  • Overall length of 2 in.
  • Constructed from tool steel (Rc 60/65)
  • Finish of 10 micro or better
  • Minus tolerance direction
  • Reversible style
  • Includes certificate of accuracy with NIST traceability