Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Tool Steel, 0.6380 in. Dia, 2.0000 in. OAL, 1 ea
Catalog No. 5000513838
Encompass
This Class ZZ plug gage is designed for high-precision measurement tasks, including go inspection and the setting of micrometers, verniers, and other measuring devices. Constructed from durable tool steel with a 60 to 65 Rc hardness, it provides long-lasting reliability in demanding industrial environments.
  • Manufactured to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microinch surface finish for precision
  • Tolerance of 0.0002 in.
  • Overall length of 2.0000 in.
Catalog No. 50-005-13838 Supplier MSC Supplier No. 88963806
This Class ZZ plug gage is designed for high-precision measurement tasks, including go inspection and the setting of micrometers, verniers, and other measuring devices. Constructed from durable tool steel with a 60 to 65 Rc hardness, it provides long-lasting reliability in demanding industrial environments.
  • Manufactured to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microinch surface finish for precision
  • Tolerance of 0.0002 in.
  • Overall length of 2.0000 in.