Learn More
MSC Vermont Gage Pin Gage, 52100 Tool Steel, 0.1105 in., 2 in. Length, 1 ea
Catalog No. 5000513855
Encompass
This Class ZZ pin gage is a precision-engineered tool designed for high-accuracy inspection, calibration of micrometers, and checking of other measuring devices. Manufactured from 52100 tool steel, this gage provides durability and wear resistance in demanding industrial environments.
  • Heat-treated to 60 to 62 RC for enhanced hardness
  • Centerless lapped and finished to 10 micro-finish or better for smooth surface quality
  • Minus tolerance of +0.0000 in./-0.0002 in. for precise dimensioning
  • Features ground ends for smooth, damage-free entry
  • Includes a certificate of accuracy with NIST traceability
  • Treated for corrosion prevention
  • Overall length of 2 in.
Catalog No. 50-005-13855 Supplier MSC Supplier No. 89011050
This Class ZZ pin gage is a precision-engineered tool designed for high-accuracy inspection, calibration of micrometers, and checking of other measuring devices. Manufactured from 52100 tool steel, this gage provides durability and wear resistance in demanding industrial environments.
  • Heat-treated to 60 to 62 RC for enhanced hardness
  • Centerless lapped and finished to 10 micro-finish or better for smooth surface quality
  • Minus tolerance of +0.0000 in./-0.0002 in. for precise dimensioning
  • Features ground ends for smooth, damage-free entry
  • Includes a certificate of accuracy with NIST traceability
  • Treated for corrosion prevention
  • Overall length of 2 in.