Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.4845 in. Diameter, Tool Steel, 1 ea.
Catalog No. 5000513835
Encompass
This Class ZZ Plus plug gage is designed for high-precision measurement tasks, including go inspection and the calibration of micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it provides reliable performance for exacting industrial applications.
  • Manufactured from tool steel for enhanced durability
  • Features a 0.4845 in. diameter
  • Maintains a 0.0002 in. tolerance
  • Includes a 10 microfinish surface
  • Meets ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-13835 Supplier MSC Supplier No. 88948450
This Class ZZ Plus plug gage is designed for high-precision measurement tasks, including go inspection and the calibration of micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it provides reliable performance for exacting industrial applications.
  • Manufactured from tool steel for enhanced durability
  • Features a 0.4845 in. diameter
  • Maintains a 0.0002 in. tolerance
  • Includes a 10 microfinish surface
  • Meets ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability