Learn More
MSC Vermont Gage Class ZZ Pin Gage, Tool Steel, 0.704 in., 2 in. OAL, 1 ea.
Catalog No. 5000513837
Encompass
This Class ZZ plug gage is designed for precision inspection, including go inspection and setting micrometers, verniers, and other measuring devices. Constructed from high-quality 52100 tool steel, the gage is heat-treated to 60 to 62 RC for durability and wear resistance.
  • Manufactured to ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 microfinish or better surface finish
  • Tolerance of +0.0002 in./-0.0000 in.
  • Round within 0.0001 in.
  • Overall length of 2.0000 in.
  • Marked with size and direction of tolerance
Catalog No. 50-005-13837 Supplier MSC Supplier No. 88970405
This Class ZZ plug gage is designed for precision inspection, including go inspection and setting micrometers, verniers, and other measuring devices. Constructed from high-quality 52100 tool steel, the gage is heat-treated to 60 to 62 RC for durability and wear resistance.
  • Manufactured to ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 microfinish or better surface finish
  • Tolerance of +0.0002 in./-0.0000 in.
  • Round within 0.0001 in.
  • Overall length of 2.0000 in.
  • Marked with size and direction of tolerance