Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Tool Steel, 0.7610 in. Diameter, 2.0000 in. Length, 1 ea.
Catalog No. 5000513689
Encompass
This Class ZZ plug gage is designed for high-precision measurement tasks, including go inspection and the setting of micrometers, verniers, and other precision measuring instruments. Constructed from 52100 tool steel and heat-treated to 60 to 65 Rc, this gage offers excellent durability and wear resistance in demanding industrial environments.
  • Diameter of 0.7610 in. with a plus tolerance of 0.0000 to +0.0002 in.
  • Overall length of 2.0000 in.
  • Finish of 10 microinch or better for surface quality.
  • Round within 0.0001 in.
  • Includes certificate of accuracy with NIST traceability.
  • Manufactured in the United States according to ASME B89.1 standards.
Catalog No. 50-005-13689 Supplier MSC Supplier No. 88976105
This Class ZZ plug gage is designed for high-precision measurement tasks, including go inspection and the setting of micrometers, verniers, and other precision measuring instruments. Constructed from 52100 tool steel and heat-treated to 60 to 65 Rc, this gage offers excellent durability and wear resistance in demanding industrial environments.
  • Diameter of 0.7610 in. with a plus tolerance of 0.0000 to +0.0002 in.
  • Overall length of 2.0000 in.
  • Finish of 10 microinch or better for surface quality.
  • Round within 0.0001 in.
  • Includes certificate of accuracy with NIST traceability.
  • Manufactured in the United States according to ASME B89.1 standards.