Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.9750 in. Diameter, 2.0000 in. Length, 1 ea.
Catalog No. 5000513694
Encompass
This Class ZZ Plus plug gage is designed for precise inspection and measurement applications. It is engineered for use in go inspection tasks, as well as for setting micrometers, verniers, and other measuring instruments to ensure dimensional accuracy. Constructed from durable tool steel, this gage meets ASME B89.1 standards for performance and reliability.
  • Manufactured from tool steel
  • Designed for go inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Features a 0.9750 in. diameter
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
  • Coating finish of 10 micro
Catalog No. 50-005-13694 Supplier MSC Supplier No. 88997507
This Class ZZ Plus plug gage is designed for precise inspection and measurement applications. It is engineered for use in go inspection tasks, as well as for setting micrometers, verniers, and other measuring instruments to ensure dimensional accuracy. Constructed from durable tool steel, this gage meets ASME B89.1 standards for performance and reliability.
  • Manufactured from tool steel
  • Designed for go inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Features a 0.9750 in. diameter
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
  • Coating finish of 10 micro