Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, Steel, 0.7470 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000511925
Encompass
This Class ZZ minus plug and pin gage is designed for high-precision inspection and calibration tasks. Manufactured from high-quality steel with a surface finish of 10 microinches, this gage provides the reliability needed for accurate measurement setups.
  • Designed for NoGo inspection, setting micrometers, verniers, and other measuring devices
  • Accuracy of 0.0002 in.
  • Overall length of 2 in.
  • Hardened to 60/65 Rc
  • Round within 0.0001 in.
  • Includes certificate of accuracy with NIST traceability
  • Size and direction of tolerance marked on the gage
Catalog No. 50-005-11925 Supplier MSC Supplier No. 89074702
This Class ZZ minus plug and pin gage is designed for high-precision inspection and calibration tasks. Manufactured from high-quality steel with a surface finish of 10 microinches, this gage provides the reliability needed for accurate measurement setups.
  • Designed for NoGo inspection, setting micrometers, verniers, and other measuring devices
  • Accuracy of 0.0002 in.
  • Overall length of 2 in.
  • Hardened to 60/65 Rc
  • Round within 0.0001 in.
  • Includes certificate of accuracy with NIST traceability
  • Size and direction of tolerance marked on the gage