Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Tool Steel, 0.0280 in. Diameter, 2.0000 in. Length, 1 ea.
Catalog No. 5000511790
Encompass
This Class ZZ Plus plug gage is designed for precision inspection tasks and measuring device calibration. Constructed from durable tool steel, it provides reliable performance for go inspection, setting micrometers, and checking verniers.
  • Manufactured from tool steel
  • Hardness rating of 60/65 Rc
  • Surface finish of 10 microinch or better
  • Roundness within .0001 in. (0.0025 mm)
  • Tolerance of .0002 in. (0.005 mm)
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-11790 Supplier MSC Supplier No. 88902804
This Class ZZ Plus plug gage is designed for precision inspection tasks and measuring device calibration. Constructed from durable tool steel, it provides reliable performance for go inspection, setting micrometers, and checking verniers.
  • Manufactured from tool steel
  • Hardness rating of 60/65 Rc
  • Surface finish of 10 microinch or better
  • Roundness within .0001 in. (0.0025 mm)
  • Tolerance of .0002 in. (0.005 mm)
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards