Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 0.2440 in., Class ZZ, 1 ea.
Catalog No. 5000511545
Encompass
This pin gage is designed for precise measurement of hole sizes and features a diameter of 0.2440 in. Constructed from 52100 tool steel and heat-treated to 60-65 RC, this instrument provides durability and consistent performance in demanding metrology applications.
  • Class ZZ accuracy with a tolerance of +0.0002/-0.0000 in.
  • Overall length of 2 in.
  • Features a 10 microfinish surface for enhanced measurement accuracy.
  • Includes a certificate of accuracy with NIST traceability to meet quality standards.
  • Manufactured in accordance with ASME B89.1 specifications.
  • Reversible style design.
Catalog No. 50-005-11545 Supplier MSC Supplier No. 88924402
This pin gage is designed for precise measurement of hole sizes and features a diameter of 0.2440 in. Constructed from 52100 tool steel and heat-treated to 60-65 RC, this instrument provides durability and consistent performance in demanding metrology applications.
  • Class ZZ accuracy with a tolerance of +0.0002/-0.0000 in.
  • Overall length of 2 in.
  • Features a 10 microfinish surface for enhanced measurement accuracy.
  • Includes a certificate of accuracy with NIST traceability to meet quality standards.
  • Manufactured in accordance with ASME B89.1 specifications.
  • Reversible style design.