Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Minus, 0.6160 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000511388
Encompass
This Class ZZ minus pin gage is designed for high-precision measurement applications, such as setting micrometers and verniers, or for use in NoGo inspections. Constructed from durable tool steel, the gage is engineered for reliable performance in demanding environments.
  • Diameter of 0.6160 in.
  • Overall length of 2 in.
  • Tolerance of +0.0000 / -0.0002 in.
  • Features a high-quality finish of 10 micro or better
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-11388 Supplier MSC Supplier No. 89061600
This Class ZZ minus pin gage is designed for high-precision measurement applications, such as setting micrometers and verniers, or for use in NoGo inspections. Constructed from durable tool steel, the gage is engineered for reliable performance in demanding environments.
  • Diameter of 0.6160 in.
  • Overall length of 2 in.
  • Tolerance of +0.0000 / -0.0002 in.
  • Features a high-quality finish of 10 micro or better
  • Includes a certificate of accuracy with NIST traceability