Learn More
MSC Vermont Gage Black Guard Oxide Pin Gage Set, Class ZZ, .001 in. Increments, 1 ea.
Catalog No. 5000515908
Encompass
These pin gage sets feature a Black Guard oxide treatment designed to resist corrosion and provide a visual indicator for wear to help save time and maintenance costs. These Class ZZ gages are suitable for a wide range of measurement applications, including determining hole sizes, calculating distances between holes, gaging slot widths, checking locations, and serving as Go/No-Go gages.
  • Features Black Guard oxide treatment for improved corrosion resistance and wear monitoring
  • Suitable for checking hole locations and slot widths
  • Compatible with Go/No-Go gaging applications
  • Includes certificate of accuracy
  • Precision increments of 0.001 in.
Catalog No. 50-005-15908 Supplier MSC Supplier No. 89990626
These pin gage sets feature a Black Guard oxide treatment designed to resist corrosion and provide a visual indicator for wear to help save time and maintenance costs. These Class ZZ gages are suitable for a wide range of measurement applications, including determining hole sizes, calculating distances between holes, gaging slot widths, checking locations, and serving as Go/No-Go gages.
  • Features Black Guard oxide treatment for improved corrosion resistance and wear monitoring
  • Suitable for checking hole locations and slot widths
  • Compatible with Go/No-Go gaging applications
  • Includes certificate of accuracy
  • Precision increments of 0.001 in.