Learn More
Cole-Parmer Troemner 311-01S Economical S.S. Class 7 Weight Set; 100 g to 1 mg
Catalog No. NC3792158 Shop All Cole-Parmer Products
Encompass_Preferred
This economical set is ideal for general-purpose applications. Weight sets are designed to achieve any total mass by combining smaller masses. All weighs feaature a satin finish surface and are marked with their nominal mass value.
Catalog No. NC3792158 Supplier Cole-Parmer Supplier No. 0102893
This economical set is ideal for general-purpose applications. Weight sets are designed to achieve any total mass by combining smaller masses. All weighs feaature a satin finish surface and are marked with their nominal mass value.