Learn More
TA Instruments Classic Aluminum Pans, DSC Sample Pans, Higher Sidewalls, Wide Variety of Samples, Use with Standard Aluminum Lids, Crimp Seal Mode, Requires Tzero Sample Press or Standard Press, Reproducible Thermal Results, 200 Pack
Catalog No. 5000580677 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Classic Aluminum Pans are DSC sample pans with higher sidewalls for analysis of a wide variety of samples. For use with Standard Aluminum Lids in crimp seal mode. Sample encapsulation requires either the Tzero Sample Press or Standard Press. These pans are versatile for general DSC applications and provide reproducible thermal results.
Catalog No. 50-005-80677 Supplier TA Instruments Supplier No. 901671.901
Waters TA Instruments Classic Aluminum Pans are DSC sample pans with higher sidewalls for analysis of a wide variety of samples. For use with Standard Aluminum Lids in crimp seal mode. Sample encapsulation requires either the Tzero Sample Press or Standard Press. These pans are versatile for general DSC applications and provide reproducible thermal results.