Learn More
Euthanex Classic Anesthesia Machine with New TEC 3 Vaporizer - Key Fill
Catalog No. 501540162
Encompass_Preferred
Classic Anesthesia Machine with New TEC 3 Vaporizer with Key Fill well. Incorporates a manual 5-output manifold and includes a built-in flow meter configuration and TEC 3 vaporizer. Select Isoflurane or Sevoflurane with purchase.
Catalog No. 50-154-0162 Supplier Euthanex Supplier No. EZ150CKF
Classic Anesthesia Machine with New TEC 3 Vaporizer with Key Fill well. Incorporates a manual 5-output manifold and includes a built-in flow meter configuration and TEC 3 vaporizer. Select Isoflurane or Sevoflurane with purchase.