Learn More
MSC PRO-SOURCE Medium Duty Trash Bags, 45 gal, 0.6 mil, 250/Pk.
Catalog No. 502866532
Encompass
PRO-SOURCE Medium Duty Trash Bags are designed for everyday waste disposal. These clear bags offer a 45 gal. capacity and a 0.6 mil thickness, making them suitable for various settings requiring a robust yet clear waste solution.
  • Made from low density resin
  • Tie closure for secure sealing
  • Coreless roll dispensing for convenience
  • Unscented for sensitive environments
  • Do not contain recycled material
  • Not FDA approved
  • Measures 40 in wide and 46 in high
Catalog No. 50-286-6532 Supplier MSC Supplier No. 56554462
PRO-SOURCE Medium Duty Trash Bags are designed for everyday waste disposal. These clear bags offer a 45 gal. capacity and a 0.6 mil thickness, making them suitable for various settings requiring a robust yet clear waste solution.
  • Made from low density resin
  • Tie closure for secure sealing
  • Coreless roll dispensing for convenience
  • Unscented for sensitive environments
  • Do not contain recycled material
  • Not FDA approved
  • Measures 40 in wide and 46 in high