Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Classic Human Skull Model with Opened Lower Jaw
GREENER_CHOICESDPSCI_ED

Catalog No. S00244 Shop All 3B Scientific Products
Change view
Click to view available options
Dimensions (L x W x H):
20 x 13.5 x 15.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S00244 20 x 13.5 x 15.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00244 Supplier 3B Scientific™ Supplier No. 1020166
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3B dental skull with opened mandible exposes tooth roots with vessels and nerves. The skull model also features the numbering of the cranial bones, bone components, fissures, foramina and other structures. This skull is a great tool for dental students and teachers alike.

  • This dental skull with opened mandible exposes tooth roots with vessels and nerves. The skull model also features the numbering of the cranial bones, bone components, fissures, foramina and other structures. The cranial sutures are shown in color, as are the meningeal vessels and venous sinuses.
    • High-quality original casts
    • Skull is handmade of hard, unbreakable plastic
    • Highly accurate representation of the fissures, foramina, processes, sutures etc.
    • Can be disassembled into Skull Cap, Base of Skull and Mandible
    • As an option, you can insert a 5-part brain (C 18) into the skull
  • This unique model of the classic human skull is useful for students and practicing doctors alike, being a great tool for learning and teaching. Not only are important anatomical features of the skull depicted but also the open jaw gives a distinctive view of teeth roots.
  • Every original 3B Scientific anatomy model now includes these additional FREE features:
    • Free access to the anatomy course 3B Smart Anatomy, hosted inside the award-winning Complete Anatomy app by 3D4Medical
    • The 3B Smart Anatomy course includes 23 digital anatomy lectures, 117 different virtual anatomy models and 39 anatomy quizzes to test your knowledge
    • Bonus: FREE warranty upgrade from 3 to 5 years with every product registration
    • Tip: You will also receive access to a free 3-day trial to all premium features of the Complete Anatomy app when you sign up for your 3B Smart Anatomy course.

Specifications

Dimensions (L x W x H) 20 x 13.5 x 15.5 cm
For Use With (Application) Learning and Teaching
Includes Free Access to the Anatomy Course 3B Smart Anatomy
Weight (Metric) 0.684 kg
Product Type Dental Skull
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.