Learn More
MARSHALL SCIENTIFIC Gilson Classic Large Volume Kit
SDP
Catalog No. NC2888824
Encompass_Preferred
The PIPETMAN Classic line is one of the world standards in pipetting and offers a wide range of continuously adjustable, air-displacement pipettes with legendary accuracy and precision. The PIPETMAN Classic Large-Volume Kit provides two pipettes that cover large volume transfers from 500 uL to 10 mL, along with appropriate filters. Made of highly resistant materials like PVDF and stainless steel, PIPETMAN Classic offers uncontested durability and reliability. Built-to-last, it is the economical solution for years of pipetting.
Catalog No. NC2888824 Supplier MARSHALL SCIENTIFIC Supplier No. F167400
The PIPETMAN Classic line is one of the world standards in pipetting and offers a wide range of continuously adjustable, air-displacement pipettes with legendary accuracy and precision. The PIPETMAN Classic Large-Volume Kit provides two pipettes that cover large volume transfers from 500 uL to 10 mL, along with appropriate filters. Made of highly resistant materials like PVDF and stainless steel, PIPETMAN Classic offers uncontested durability and reliability. Built-to-last, it is the economical solution for years of pipetting.