Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Classification of Soil Layers
SCI_ED

Catalog No. S01407
Change view
Click to view available options
Grade:
7 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01407 7 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S01407 Supplier Neo/Sci Corporation Supplier No. 204143

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover the conditions that caused your local soils to form.

  • Determine how soils are formed
  • Create a sample soil profile using the provided soil horizon cards
  • Determine the environmental changes that cause those soils to form
  • Portable and practical soil cards

Order Info

Grades 7 to 12. Class size: 40 students.

Compliance

NSES: A, D, F, G

Specifications

Grade 7 to 12
For Use With (Application) Determine how soils are formed and create a sample soil profile
Product Type Classification of Soil Layers Lab Investigation
Sufficient For 40 Students
Certifications/Compliance NSES: A, D, F, G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.